missing translation for 'onlineSavingsMsg'
Learn More

Chloromethoxysulfonyl Chloride, TRC

Product Code. 30362686
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362686 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362686 Supplier Toronto Research Chemicals Supplier No. TRCC365096500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloromethoxysulfonyl Chloride
CAS 49715-04-0
Molecular Formula C H2 Cl2 O3 S
Quantity 500 mg
Synonym Chloromethyl Chlorosulfate, Chlorosulfuric Acid, Chloromethyl Ester
InChI Formula InChI=1S/CH2Cl2O3S/c2-1-6-7(3,4)5/h1H2
SMILES ClCOS(=O)(=O)Cl
IUPAC Name chloro(chlorosulfonyloxy)methane
Molecular Weight (g/mol) 164.996
Formula Weight 163.91
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.