missing translation for 'onlineSavingsMsg'
Learn More

Chloroiodomethane, 98%, stabilized

Product Code. 10366451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366451 100 mL Glass bottle 100mL
10515401 25 mL Glass bottle 25mL
11914511 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366451 Supplier Thermo Scientific Acros Supplier No. 222151000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-71-5 | CH2ClI | 176.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-71-5 , 7440-22-4
Molecular Formula CH2ClI
Molecular Weight (g/mol) 176.38
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane
SMILES C(Cl)I

Specifications

Color Pink-Red to Yellow
Density 2.4200g/mL
Boiling Point 108.0°C to 109.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2I
Refractive Index 1.5812 to 1.5832
Quantity 100 mL
Beilstein 01,71
Fieser 10,89; 17,115
Specific Gravity 2.42
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
Stabilizer Copper
Formula Weight 176.38
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloroiodomethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.