missing translation for 'onlineSavingsMsg'
Learn More

Chloroiodomethane, 98%, stab. with copper

Code produit 11384559
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
500 g
Conditionnement:
10g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11384559 10 g 10g
11394559 50 g 50g
11304569 250 g 250g
11498863 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11384559 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur A11032.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 593-71-5 | CH2ClI | 176.381 g/mol

Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

Solubility
Miscible with acetone, benzene, diethyl ether and alcohol.

Notes
It is incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 593-71-5
Molecular Formula CH2ClI
MDL Number MFCD00001078
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane

Spécification

Density 2.416
Boiling Point 108°C to 109°C
Flash Point None
Refractive Index 1.5822
Quantity 10 g
UN Number UN2810
Beilstein 1730802
Sensitivity Light sensitive
Solubility Information Miscible with acetone,benzene,diethyl ether and alcohol.
SMILES C(Cl)I
Molecular Weight (g/mol) 176.381
Formula Weight 176.38
Percent Purity 98%
Chemical Name or Material Chloroiodomethane, Stabilized with copper
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Chloroiodomethane
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informations supplémentaires
  • MIXTURE LIST-Contain: Chloroiodomethane

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.