missing translation for 'onlineSavingsMsg'
Learn More

Chloroiodomethane, 98%, stab. with copper

Product Code. 11394559
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
500 g
Unit Size:
10g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394559 50 g 50g
11384559 10 g 10g
11304569 250 g 250g
11498863 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11394559 Supplier Thermo Scientific Alfa Aesar Supplier No. A11032.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-71-5 | CH2ClI | 176.381 g/mol

Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

Solubility
Miscible with acetone, benzene, diethyl ether and alcohol.

Notes
It is incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-71-5
Molecular Formula CH2ClI
MDL Number MFCD00001078
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane

Specifications

Density 2.416
Boiling Point 108°C to 109°C
Flash Point None
Refractive Index 1.5822
Quantity 50 g
UN Number UN2810
Beilstein 1730802
Sensitivity Light sensitive
Solubility Information Miscible with acetone,benzene,diethyl ether and alcohol.
SMILES C(Cl)I
Molecular Weight (g/mol) 176.381
Formula Weight 176.38
Percent Purity 98%
Chemical Name or Material Chloroiodomethane, Stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.