missing translation for 'onlineSavingsMsg'
Learn More

Chlorogenic Acid, TRC

Product Code. 17989744
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17989744 1 g 1g
17919924 250 mg 250mg
17958114 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17989744 Supplier Toronto Research Chemicals Supplier No. TRCC3665401G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-97-9
Molecular Formula C16 H18 O9
Molecular Weight (g/mol) 354.31
Synonym Chlorogenic acid (8CI), Cyclohexanecarboxylic acid, 3-[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxy-, (1S,3R,4R,5R)- (9CI), Cyclohexanecarboxylic acid, 3-[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxy-, [1S-(1α,3β,4α,5α)]-, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, 3-Caffeoylquinic acid, 3-O-(3,4-Dihydroxycinnamoyl)-D-quinic acid, 3-O-Caffeoylquinic acid, Caffeoylquinic acid, Heriguard, NSC 407296, NSC 70861, SW 85834
IUPAC Name (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@@H]1O)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H18O9/c17-9-3-1-8(5-10(9)18)2-4-13(20)25-12-7-16(24,15(22)23)6-11(19)14(12)21/h1-5,11-12,14,17-19,21,24H,6-7H2,(H,22,23)/b4-2+/t11-,12-,14-,16+/m1/s1
Formula Weight 354.0951
Percent Purity >95%
Chemical Name or Material Chlorogenic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.