missing translation for 'onlineSavingsMsg'
Learn More

Chlorogenic acid, MedChemExpress

Product Code. 16412346
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412346 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412346 Supplier Medchem Express Supplier No. HYN0055
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chlorogenic acid is a major phenolic compound in coffee and tea. It plays several important and therapeutic roles such as antioxidant activity, antibacterial, hepatoprotective, cardioprotective, anti-inflammatory, antipyretic, neuroprotective, anti-obesity, antiviral, anti-microbial, anti-hypertension.

Chlorogenic acid is a major phenolic compound in coffee and tea. It plays several important and therapeutic roles such as antioxidant activity, antibacterial, hepatoprotective, cardioprotective, anti-inflammatory, antipyretic, neuroprotective, anti-obesity, antiviral, anti-microbial, anti-hypertension.

Specifications

Chemical Name or Material Chlorogenic acid
Physical Form Powder
CAS 327-97-9
Color Off-White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C16H18O9
For Use With (Application) COVID-19-anti-virus
Synonym 3-O-Caffeoylquinic acid Heriguard NSC-407296
Solubility Information DMSO : 100 mg/mL (282.24 mM; Need ultrasonic)
SMILES O=C([C@@]1(O)C[C@@H](OC(/C=C/C2=CC=C(O)C(O)=C2)=O)[C@H](O)[C@H](O)C1)O
Molecular Weight (g/mol) 354.31
Formula Weight 354.31
Percent Purity 98.96%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.