missing translation for 'onlineSavingsMsg'
Learn More

Chlorogenic acid, 99.45%, For HPLC analysis, MP Biomedicals™

Product Code. 11467921 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467921 1 g 1g
11477921 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467921 Supplier MP Biomedicals Supplier No. 0215061801
  • ONLINE EXCLUSIVE PRICE
    £86.10
    £80.75 / 1g
    Save £5.35
    6% Off
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 327-97-9
Molecular Formula C16H18O9
Molecular Weight (g/mol) 354.31
MDL Number MFCD00003862
InChI Key CWVRJTMFETXNAD-JUHZACGLSA-N
Synonym chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri
PubChem CID 1794427
ChEBI CHEBI:16112
IUPAC Name (1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O

Specifications

Color White
Assay Percent Range 99.45%
Quantity 1 g
Formula Weight 354.3
Physical Form Crystalline Powder
Chemical Name or Material Chlorogenic acid, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.