missing translation for 'onlineSavingsMsg'
Learn More

Chloroformic Acid Chloromethyl Ester, TRC

Product Code. 18041000
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18041000 50 g 50g
18022710 25 g 25g
18081000 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18041000 Supplier Toronto Research Chemicals Supplier No. TRCC36650050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22128-62-7
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.94
Synonym Chloro-formic acid Chloromethyl Ester, Chloromethoxycarbonyl Chloride, Chloromethyl Carbonochloridate, Chloromethyl Chlorocarbonate, Chloromethyl Chloroformate
IUPAC Name chloromethyl carbonochloridate
SMILES C(OC(=O)Cl)Cl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C2H2Cl2O2/c3-1-6-2(4)5/h1H2
Chemical Name or Material Chloroformic Acid Chloromethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.