missing translation for 'onlineSavingsMsg'
Learn More

Chloroform (STB ETOH), BAKER ANALYZED Pesticide Reagent, J.T. Baker™

Product Code. 15508334 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15508334 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15508334 Supplier JT Baker Supplier No. 5285.2500
  • ONLINE EXCLUSIVE PRICE
    £478.19
    £411.30 / 2.5 L
    Save £66.89
    13% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specially developed for use in evaluation of pesticide and insecticide residues in food, feed, water or soil samples. Chloroform (STB ETOH), BAKER ANALYZED Pesticide Reagent, J.T. Baker™ is suitable for all analysis of common pesticides.

J.T. Baker BAKER ANALYZED Pesticide Reagent Solvents:

  • Suitable for analysis of most common pesticides, such as 2-chlorobiphenyl , Heptachlor, Aldrin, Parathion, Dieldrin, Endrin, DDT, Dioxine, etc.
  • Specified to the required low residue levels (5 ppm) for use in research and quality control laboratories
  • Ideal for gas chromatography applications, testing for ECD and/or FID sensitive impurities, organic residues, and residue after evaporation

Specifications

Melting Point -63°C
CAS 67-66-3
Color Colorless
Acidity Max. 0.0005meq/g
Density 1.5g/cm3 at 20°C
Quantity 2.5 L
Ignition Point 982°C
Odor Sweetish
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 5 ppm
Molecular Formula CHCl3
MDL Number MFCD00000826
Viscosity 0.56 mPas at 20°C
Water Max. 0.05%
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Boiling Point 62°C
Solubility Information 8g/L in water
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name trichloromethane
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.37g/mol
Percent Purity Min. 99.8%
Grade Pesticide
Chemical Name or Material Chloroform
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.