missing translation for 'onlineSavingsMsg'
Learn More

Chloroform (Stabilized with ~1% Ethanol), TRC

Product Code. 30364400
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364400 250 mL -
30359184 100 mL 100mL
30358845 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364400 Supplier Toronto Research Chemicals Supplier No. TRCC366490250ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula C H Cl3
Molecular Weight (g/mol) 119.38
Synonym Methane, trichloro-, Chloroform (8CI), Trichloromethane, F 20, HCC 20, NSC 77361, R 20, R 20 (refrigerant), Trichloroform
IUPAC Name chloroform
SMILES ClC(Cl)Cl

Specifications

Quantity 250 mL
InChI Formula InChI=1S/CHCl3/c2-1(3)4/h1H
Formula Weight 117.9144
Chemical Name or Material Chloroform
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.