missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.0-99.4% (GC), Solstice
BIOPHARMA

Product Code. 15653700
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15653700 4 x 2.5 L Glass Bottle Pack of 4
15643700 1 L Glass Bottle 1 L
15675705 2.5 L Glass Bottle 2.5 L
15775123 2.5 L Glass Bottle 1 L
15663700 6 x 1 L Glass Bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15653700 Supplier Honeywell-Riedel-de Haen Supplier No. 322114X2.5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.0-99.4% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3 , 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C (literature)
CAS Min % ≥90.0000%, ≥0.1000%
CAS Max % ≤100.0000%, <1.0000%
Density 1.476-1.483 g/mL at 20 °C; 1.492 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Packaging Glass Bottle
Linear Formula CHCl3
UN Number UN1888
Beilstein 1731042
Boiling Point 60°C to 62°C
Grade puriss. p.a.
Chemical Name or Material Chloroform, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.0-99.4% (GC)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.