Learn More
Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene, Fisher Chemical™
Brand: Fisher Chemical C/4962/17
28.50 GBP valid until 2022-05-29
Use promo code "17501" to get your promotional price.
Code : 25
Additional Details : CAS Number : 67-66-3 Weight : 3.72500kg Transport : UN number : 1888 Chem class : 6.1 Pack group : III
Quantity | 2.5L |
---|
Description
Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene
Chemical Identifiers
67-66-3 | |
Colorless | |
-63°C | |
826 | |
Amber glass bottle | |
0.56 mPaS at 20°C |
61°C | |
119.38g/mol | |
Liquid | |
CHCl3 | |
213mbar at 20°C |
Specifications
67-66-3 | |
826 | |
Liquid | |
Colorless | |
2.5L | |
213mbar at 20°C |
CHCl3 | |
119.38g/mol | |
61°C | |
-63°C | |
Amber glass bottle | |
0.56 mPaS at 20°C |
Safety and Handling
- Chloroform, stabilized with amylene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H372-Causes damage to organs.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .