missing translation for 'onlineSavingsMsg'
Learn More

Chloroform-d, for NMR, 100.0 atom % D, stabilized with silver

Product Code. 16308206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16308206 25 mL 25mL
16318206 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16308206 Supplier Thermo Scientific Acros Supplier No. 464050250
  • ONLINE EXCLUSIVE PRICE
    £116.00
    £112.00 / 25mL
    Save £4.00
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 865-49-6 | CHCl3 | 120.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-49-6
Molecular Formula CHCl3
Molecular Weight (g/mol) 120.38
InChI Key HEDRZPFGACZZDS-MICDWDOJSA-N
IUPAC Name trichloro(²H)methane
SMILES [2H]C(Cl)(Cl)Cl

Specifications

Density 1.5000 g/mL
Specific Gravity 1.5
Quantity 25 mL
Melting Point -64.0°C
Boiling Point 60.0°C
Linear Formula CDCl3
Solubility Information Solubility aq. soln.: 8.2 g/L (20°C).
Formula Weight 120.39
Grade NMR
Chemical Name or Material Chloroform-d

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.