Product Code. 15652770
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
15652770
1 L
Glass Bottle
1 L
15662770
2.5 L
Glass Bottle
2.5 L
15682770
6 x 1 L
Glass Bottle
Pack of 6
15672770
4 x 2.5 L
Glass Bottle
Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code.
15652770
Supplier
Honeywell-Chromasolv
Supplier No.
256691L
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer,≥99.8%
For gas chromatography (GC) Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
CAS
67-66-3
,
64-17-5
Molecular Formula
CHCl3
Molecular Weight (g/mol)
119.37
MDL Number
MFCD00000826
InChI Key
HEDRZPFGACZZDS-UHFFFAOYSA-N
PubChem CID
6212
ChEBI
CHEBI:35255
IUPAC Name
trichloromethane
SMILES
ClC(Cl)Cl
Specifications
Melting Point
-63°C (literature)
CAS Min %
≥90.0000%, ≥0.1000%
CAS Max %
≤100.0000%, <1.0000%
Density
1.492 g/mL at 25 °C(lit.)
Boiling Point
60°C to 62°C
Quantity
1 L
Packaging
Glass Bottle
Linear Formula
CHCl3
UN Number
UN1888
Beilstein
1731042
Synonym
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Percent Purity
>99.8%
Grade
Residue Analysis
Assay
GC product+ethanol
Chemical Name or Material
Chloroform, CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer, ≥99.8%
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?