missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, CHROMASOLV™ Plus, ≥99.9%, Amylenes as stabilizer, Honeywell™

Product Code. 15606120
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15606120 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15606120 Supplier Honeywell-Chromasolv Supplier No. 6504981L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 67-66-3
Density 1.492g/mL at 25°C, 1.48g/mL at 25°C
Quantity 1 L
Molecular Formula CHCl3
Vapor Pressure 160 mmHg (20°C)
MDL Number MFCD00000826
UN Number UN1888
Beilstein 1731042
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name trichloromethane
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.38g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Plus
Chemical Name or Material Chloroform
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: chloroform
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.