missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Biotech Grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer, Honeywell Burdick & Jackson™

Product Code. 15625690
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15625690 2 L Glass Bottle 2 L
15615690 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15625690 Supplier Honeywell-Burdick and Jackson Supplier No. 4961892L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Biotech Grade,≥99.8%, contains 0.5-1.0% ethanol as stabilizer

  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 1.48 g/mL at 25 °C; 1.492 g/mL at 25 °C(lit.)
Quantity 2 L
Packaging Glass Bottle
Linear Formula CH3CH2OH
UN Number UN1888
Beilstein 1731042
Boiling Point 61°C
Formula Weight 119.38
Percent Purity ≥99.8%
Grade Biotech
Chemical Name or Material Chloroform, Biotech Grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.