missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, 99%, stab. with 0.8-1% ethanol, Thermo Scientific Chemicals

Product Code. 11429830
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL

Chloroform is a commonly used laboratory solvent. It was previously used as an anesthetic. It is also used as a precursor to PTFE and refrigerants. Chloroform was used in the past as an extraction solvent for fats, oils and greases. It is useful for the rapid and simple release of periplasmic proteins. The mixture of guanidinium thiocyanate-phenol-chloroform, plays an essential role in the RNA isolation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroform is a commonly used laboratory solvent. It was previously used as an anesthetic. It is also used as a precursor to PTFE and refrigerants. Chloroform was used in the past as an extraction solvent for fats, oils and greases. It is useful for the rapid and simple release of periplasmic proteins. The mixture of guanidinium thiocyanate-phenol-chloroform, plays an essential role in the RNA isolation process.

Solubility
Miscible with diethyl ether, oils, ligroin, alcohol, acetone, carbon tetrachloride and carbondisulfide.Slightly miscible with water.

Notes
It is a stable and light sensitive chemical compound. Incompatible with metals, alkalies. Keep the material away from heat, sparks and flame and other sources of ignition. Store it in a cool, dry, well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Density 1.492
Boiling Point 60.5°C to 61.5°C
Quantity 2500 mL
Odor Characteristic
Refractive Index 1.444
UN Number UN1888
Beilstein 1731042
Merck Index 14,2141
Solubility Information Miscible with diethyl ether,oils,ligroin,alcohol,acetone,carbon tetrachloride and carbondisulfide.Slightly miscible with water.
IUPAC Name trichloromethane
Formula Weight 119.38
Percent Purity 99%
Chemical Name or Material Chloroform, stabilized with 0.8 to 1% ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'