missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, 99+%, Extra Pure, Stabilised with Amylene, SLR, Fisher Chemical™

Product Code. p-8015118 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10488400

Brand: Fisher Chemical C/4920/08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Color Colorless
Boiling Point 61°C
Quantity 500 mL
Packaging Amber Glass Bottle
Physical Form Liquid
MDL Number 826
Vapor Pressure 213mbar at 20°C
Viscosity 0.56 mPaS at 20°C
IUPAC Name trichloromethane
Formula Weight 119.38g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.