missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
70.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
12.50 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
Use promo code "22914" to get your promotional price.
12.50 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
CHCl3, CAS Number-67-66-3, 2.5L, 61 deg.C, Colorless, 119.38g/mol, -63 deg.C, 826, Plastic Coated Glass bottle, Liquid, 213mbar at 20 deg.C, 0.56 mPaS at 20 deg.C
Specifications
Melting Point | -63°C |
Color | Colorless |
Boiling Point | 61°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
MDL Number | MFCD00000826 |
Vapor Pressure | 213mbar at 20°C |
Viscosity | 0.56 mPaS at 20°C |
IUPAC Name | chloroform |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'