missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
42.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Beschreibung
CHCl3, CAS Number-67-66-3, 2.5L, 61 deg.C, Colorless, 119.38g/mol, -63 deg.C, 826, Plastic Coated Glass bottle, Liquid, 213mbar at 20 deg.C, 0.56 mPaS at 20 deg.C
Spezifikation
Melting Point | -63°C |
Color | Colorless |
Boiling Point | 61°C |
Quantity | 1 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
MDL Number | 826 |
Vapor Pressure | 213mbar at 20°C |
Viscosity | 0.56 mPaS at 20°C |
IUPAC Name | trichloromethane |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Chloroform, stabilized with amylene
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Chloroform, 1-Pentene
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'