missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Chloroform

Product Code. 15915450
Click to view available options
Quantity:
7 L
18 L
Unit Size:
18 L
7 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15915450

Brand: Riedel-de Haën 3221118L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chloroform, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.0 to 99.4% (GC).

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym Methylidyne trichloride, Trichloromethane
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

pH No data available
Melting Point -63°C
CAS Min % ≥90.00%
Color Colorless
CAS Max % ≤100.00%
Density 1.48 g/cm3 (20°C)
Boiling Point 60°C to 62°C (1.013 hPa)
Quantity 18 L
Flash Point Not applicable
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss. p.a.
Linear Formula CHCl3
Vapor Pressure 210 hPa (20°C)
UN Number UN1888
Viscosity No data available
Beilstein 1731042
Percent Purity 99.00 to 99.40%
Grade Reagent
Chemical Name or Material Chloroform
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.