missing translation for 'onlineSavingsMsg'
Learn More

Solstice Chloroform

Product Code. 16695733 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
7 L
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16695733 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 16695733 Supplier Solstice Supplier No. 1329507LRC
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Reagent Grade Chloroform, contains 0.5 to 1.0% ethanol as stabilizer, ≥99.8%.

  • ≥95% for general laboratory use.

Specifications

pH No data available
Melting Point -61.3°C
CAS 67-66-3
Color Colorless
CAS Max % <100.00%
Density 1.485 g/cm3 (20°C)
Quantity 7 L
Flash Point Not applicable
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Molecular Formula CHCl3
Linear Formula CHCl3
Vapor Pressure 211 hPa (20°C)
MDL Number MFCD00000826
Viscosity 0.56 mPa.s (20°C)
UN Number UN1888
Beilstein 1731042
Synonym Methylidyne trichloride, Trichloromethane
Boiling Point 61°C (1.013 hPa)
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name trichloromethane
Molecular Weight (g/mol) 119.37
Percent Purity ≥99.80%
Grade Reagent
Chemical Name or Material Chloroform
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.