missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorodiphenylphosphine, TRC
Click to view available options
Quantity:
5 g
Specifications
Specifications
| Chemical Name or Material | Chlorodiphenylphosphine |
| CAS | 1079-66-9 |
| Molecular Formula | C12H10ClP |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C12H10ClP/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H |
| SMILES | ClP(c1ccccc1)c2ccccc2 |
| IUPAC Name | chloro(diphenyl)phosphane |
| Molecular Weight (g/mol) | 220.63 |
| Formula Weight | 220.0209 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction