missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylphosphine, 98+%

Product Code. 11374058
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374058 10 g 10g
11384058 50 g 50g
11364058 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11374058 Supplier Thermo Scientific Alfa Aesar Supplier No. 041352.09
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1079-66-9 | C12H10ClP | 220.64 g/mol

Chlorodiphenylphosphine is used in the synthesis of various phosphines and an useful reagent in for the introduction of Ph2P group. It is also useful as an intermediate to make antioxidants, flame retardants, stabilizers, catalysts, photoinitiators and optical brighteners. It is also used to produce diphenylphosphinous acid methyl esters. It is used as a precursor in the preparation of various phosphines. It is involved in the preparation of phosphines, which is used as stabilizers for plastics, flame retardants and UV-hardening paint systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1079-66-9
Molecular Formula C12H10ClP
Molecular Weight (g/mol) 220.64
MDL Number MFCD00000529
InChI Key XGRJZXREYAXTGV-UHFFFAOYSA-N
Synonym chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
PubChem CID 66180
IUPAC Name chloro(diphenyl)phosphane
SMILES ClP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 14°C to 16°C
Color Colorless to Yellow
Density 1.204 g/mL
Boiling Point 318°C to 321°C
Flash Point 138°C (280°F)
Odor Acrid
Linear Formula (C6H5)2PCl
Refractive Index 1.636
Quantity 10 g
UN Number UN3265
Beilstein 512032
Sensitivity Air and moisture sensitive
Solubility Information Miscible with alcohol. Slightly miscible with ammonia.
Formula Weight 220.64
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Chlorodiphenylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.