missing translation for 'onlineSavingsMsg'
Learn More
Learn More
chlorodiphenylphosphine, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.2290g/mL |
| Boiling Point | 320.0°C |
| Flash Point | 138°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | (C6H5)2PCl |
| Refractive Index | 1.6340 to 1.6370 |
| Quantity | 500 g |
| Show More |
Safety and Handling
Product Identifier
- Chlorodiphenylphosphine
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction