missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylmethane, TRC

Product Code. 30352379
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30352379 50 g
30355346 100 g
30353295 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30352379 Supplier Toronto Research Chemicals Supplier No. TRCC36801550G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-99-3
Molecular Formula C13H11Cl
Molecular Weight (g/mol) 202.68
Synonym 1,1'-(Chloromethylene)bisbenzene, Benzhydryl Chloride, ChlorodiphenylmethaneDiphenylchloromethane, Diphenylmethyl Chloride, NSC 76584, alpha-Chlorodiphenylmethane
IUPAC Name [chloro(phenyl)methyl]benzene
SMILES ClC(c1ccccc1)c2ccccc2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C13H11Cl/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H
Formula Weight 202.0549
Chemical Name or Material Chlorodiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.