missing translation for 'onlineSavingsMsg'
Learn More

chlorodiphenylmethane, 98%

Product Code. 10177753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177753 50 mL Glass bottle 50mL
10478500 5 mL Glass bottle 5mL
10091840 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10177753 Supplier Thermo Scientific Acros Supplier No. 154660500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-99-3 | C13H11Cl | 202.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-99-3
Molecular Formula C13H11Cl
Molecular Weight (g/mol) 202.68
MDL Number MFCD00000855
InChI Key ZDVDCDLBOLSVGM-UHFFFAOYSA-N
Synonym benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene
PubChem CID 7035
IUPAC Name [chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl

Specifications

Melting Point 15°C to 17°C
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 270°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CHCl
Refractive Index 1.5940 to 1.597
Quantity 50 mL
Specific Gravity 1.14
Solubility Information Solubility in water: slightly soluble
Formula Weight 202.68
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chlorodiphenylmethane
Show More Show Less
compliance-icons
Product Identifier
  • Chlorodiphenylmethane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.