missing translation for 'onlineSavingsMsg'
Learn More

chlorodimethylthexylsilane, 95%, Thermo Scientific™

Catalog Number 10732212
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
67373-56-2
C8H19ClSi
178.78
MFCD00012175
KIGALSBMRYYLFJ-UHFFFAOYSA-N
dimethylthexylsilyl chloride, thexyldimethylchlorosilane, chloro dimethyl thexylsilane, chloro 2,3-dimethylbutan-2-yl dimethylsilane, dimethylthexylchlorosilane, thexyldimethylsilyl chloride, chlorodimethyl 1,1,2-trimethylpropyl silane, dimethylthexylsilylchloride, tds-cl, acmc-209nyu
144239
chloro(2,3-dimethylbutan-2-yl)dimethylsilane
CC(C)C(C)(C)[Si](C)(C)Cl
This item is not returnable. View return policy
67373-56-2
C8H19ClSi
178.78
MFCD00012175
KIGALSBMRYYLFJ-UHFFFAOYSA-N
dimethylthexylsilyl chloride, thexyldimethylchlorosilane, chloro dimethyl thexylsilane, chloro 2,3-dimethylbutan-2-yl dimethylsilane, dimethylthexylchlorosilane, thexyldimethylsilyl chloride, chlorodimethyl 1,1,2-trimethylpropyl silane, dimethylthexylsilylchloride, tds-cl, acmc-209nyu
144239
chloro(2,3-dimethylbutan-2-yl)dimethylsilane
CC(C)C(C)(C)[Si](C)(C)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.9000g/mL
Boiling Point 55.0°C (10.0 mmHg)
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl
Refractive Index 1.4480 to 1.4500
Quantity 25 g
Specific Gravity 0.9
Formula Weight 178.78
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material chlorodimethylthexylsilane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'