missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl(pentafluorophenyl)silane, 96%

Product Code. p-7043389
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11330104

Brand: Thermo Scientific Alfa Aesar L00550.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20082-71-7 | C8H6ClF5Si | 260.66 g/mol

It is used as a GC reagent for detection of sterols at low concentration as flophemesyl derivatives, readily detected by electron capture. Chlorodimethyl(pentafluorophenyl)silane is an extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a GC reagent for detection of sterols at low concentration as flophemesyl derivatives, readily detected by electron capture. Chlorodimethyl(pentafluorophenyl)silane is an extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20082-71-7
Molecular Formula C8H6ClF5Si
Molecular Weight (g/mol) 260.66
MDL Number MFCD00000500
InChI Key PQRFRTCWNCVQHI-UHFFFAOYSA-N
Synonym pentafluorophenyldimethylchlorosilane, flophemesyl chloride, chlorodimethylpentafluorophenylsilane, chlorodimethyl perfluorophenyl silane, silane, chlorodimethyl pentafluorophenyl, chlorodimethyl pentafluorophenyl silane, benzene, 1-chlorodimethylsilyl-2,3,4,5,6-pentafluoro, chlorodimethyl 2,3,4,5,6-pentafluorophenyl silane, chloro dimethyl pentafluorophenyl silane, dimethylpentafluorophenylchlorosilane
PubChem CID 88361
IUPAC Name chloro-dimethyl-(2,3,4,5,6-pentafluorophenyl)silane
SMILES C[Si](C)(Cl)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Boiling Point 78°C to 81°C (10mmHg)
Quantity 1 g
UN Number UN2987
Beilstein 2858407
Flash Point 95°C (203°F)
Solubility Information Reacts with water.
Formula Weight 260.66
Percent Purity 96%
Refractive Index 1.447
Sensitivity Moisture sensitive
Density 1.381
Chemical Name or Material Chlorodimethyl(pentafluorophenyl)silane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.