Få mere at vide
Chlorodimethyl(pentafluorophenyl)silane, 96%
Beskrivelse
It is used as a GC reagent for detection of sterols at low concentration as flophemesyl derivatives, readily detected by electron capture. Chlorodimethyl(pentafluorophenyl)silane is an extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a GC reagent for detection of sterols at low concentration as flophemesyl derivatives, readily detected by electron capture. Chlorodimethyl(pentafluorophenyl)silane is an extremely sensitive derivatizing agent for the analysis of sterols and lower aliphatic alcohols by electron capture gas chromatography.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Tekniske data
Tekniske data
| Boiling Point | 78°C to 81°C (10mmHg) |
| Quantity | 5 g |
| UN Number | UN2987 |
| Beilstein | 2858407 |
| Flash Point | 95°C (203°F) |
| Solubility Information | Reacts with water. |
| Formula Weight | 260.66 |
| Percent Purity | 96% |
| Refractive Index | 1.447 |
| Sensitivity | Moisture sensitive |
| Vis mere |
RUO – Research Use Only
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.