missing translation for 'onlineSavingsMsg'
Learn More

Chlorodi(p-tolyl)phosphine, 95%

Product Code. 11425640
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11425640

Brand: Thermo Scientific Alfa Aesar H25889.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1019-71-2 | C14H14ClP | 248.69 g/mol

Chlorodi(p-tolyl)phosphine is used as a reactant for synthesis of palladium catalysts for C-C bond forming cross-coupling reactions, ligands for nickel(0)-catalyzed isomerization reactions, phosphinosulfonamide nickel complexes for oligomerization reactions. It is a reactant for phosphinylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodi(p-tolyl)phosphine is used as a reactant for synthesis of palladium catalysts for C-C bond forming cross-coupling reactions, ligands for nickel(0)-catalyzed isomerization reactions, phosphinosulfonamide nickel complexes for oligomerization reactions. It is a reactant for phosphinylation reactions.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019-71-2
Molecular Formula C14H14ClP
Molecular Weight (g/mol) 248.69
MDL Number MFCD01630844
InChI Key BJBXRRHIBSXGLF-UHFFFAOYSA-N
Synonym bis 4-methylphenyl chlorophosphine, chlorodi p-tolyl phosphine, chlorodi-p-tolylphosphine, chlorobis 4-methylphenyl phosphine, bis 4-methylphenyl phosphinous chloride, chlorobis 4-methylphenyl phosphane, chloro-bis 4-methylphenyl phosphane, di-p-tolylchlorophosphine, chloro bis-p-tolyl phosphane, di p-tolyl phosphinous chloride
PubChem CID 4188608
IUPAC Name chloro-bis(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)Cl

Specifications

Density 1.159
Boiling Point 180°C to 184°C (10 mmHg)
Quantity 5 g
UN Number UN1760
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 248.69
Percent Purity 95%
Chemical Name or Material Chlorodi(p-tolyl)phosphine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.