missing translation for 'onlineSavingsMsg'
Learn More

Chlorodenafil, TRC

Product Code. 30136473
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136473 2.5 g 2.5g
17942965 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136473 Supplier Toronto Research Chemicals Supplier No. TRCC3655352.5G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1058653-74-9
Molecular Formula C19 H21 Cl N4 O3
Molecular Weight (g/mol) 388.85
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[5-(2-chloroacetyl)-2-ethoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-, Chlorodenafil
IUPAC Name 5-[5-(2-chloroacetyl)-2-ethoxyphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)C(=O)CCl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C19H21ClN4O3/c1-4-6-13-16-17(24(3)23-13)19(26)22-18(21-16)12-9-11(14(25)10-20)7-8-15(12)27-5-2/h7-9H,4-6,10H2,1-3H3,(H,21,22,26)
Formula Weight 388.1302
Percent Purity >95%
Chemical Name or Material Chlorodenafil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.