missing translation for 'onlineSavingsMsg'
Learn More

Chlorocyclohexane, 98%

Product Code. 11366896
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366896 1000 g 1kg
11356896 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11366896 Supplier Thermo Scientific Alfa Aesar Supplier No. L04349.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 542-18-7 | C6H11Cl | 118.604 g/mol

Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.

Solubility
Soluble in water.0.02 g/L (20°C). Soluble in alcohol and diethyl ether.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-18-7
Molecular Formula C6H11Cl
Molecular Weight (g/mol) 118.604
MDL Number MFCD00003822
InChI Key UNFUYWDGSFDHCW-UHFFFAOYSA-N
Synonym cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane
PubChem CID 10952
ChEBI CHEBI:39156
IUPAC Name chlorocyclohexane
SMILES C1CCC(CC1)Cl

Specifications

Melting Point -44°C
Density 1
Boiling Point 141°C to 143°C
Flash Point 28°C (82°F)
Odor Unpleasant
Refractive Index 1.462
Quantity 1000 g
UN Number UN1993
Beilstein 1900796
Merck Index 14,2732
Solubility Information Soluble in water.0.02g/L (20°C). Soluble in alcohol and diethyl ether.
Formula Weight 118.61
Percent Purity 98%
Chemical Name or Material Chlorocyclohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.