missing translation for 'onlineSavingsMsg'
Learn More

Chlorocyclobutane, 97%

Product Code. 11424264
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424264 1 g 1g
11434264 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424264 Supplier Thermo Scientific Alfa Aesar Supplier No. A12561.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1120-57-6 | C4H7Cl | 90.55 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store at 4°C. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-57-6
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00013736
InChI Key STJYMUBZVMSMBP-UHFFFAOYSA-N
Synonym cyclobutyl chloride, cyclobutane, chloro, chloro-cyclobutane, cyclobutylchloride, chloranylcyclobutane, 1-chlorocyclobutane, acmc-1butg, chlorocyclobutane, 97+%
PubChem CID 70712
IUPAC Name chlorocyclobutane
SMILES C1CC(C1)Cl

Specifications

Density 0.991
Boiling Point 80°C to 81°C
Flash Point −8°C (17°F)
Odor Slightly Sweetish
Refractive Index 1.4332
Quantity 1 g
UN Number UN1127
Beilstein 1900414
Solubility Information Insoluble in water.
Formula Weight 90.55
Percent Purity 97%
Chemical Name or Material Chlorocyclobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.