missing translation for 'onlineSavingsMsg'
Learn More

Chlorocholine Chloride, TRC

Product Code. 30138263
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138263 100 g 100g
30009371 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138263 Supplier Toronto Research Chemicals Supplier No. TRCC364945100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-81-5
Molecular Formula C5 H13 Cl N . Cl
Molecular Weight (g/mol) 158.07
Synonym Chlormequat Chloride, (2-Chloroethyl)trimethylammonium chloride (6CI,7CI), Ammonium, (2-chloroethyl)trimethyl-, chloride (8CI), Ethanaminium, 2-chloro-N,N,N-trimethyl-, chloride (9CI), (β-Chloroethyl)trimethylammonium chloride, 60CS16, Antywylegacz, Antywylegacz Plynny, Antywylegacz plynny 675SL, Barleyquat B, Bercema CCC, C5 SUN, CCC, Chlorcholine chloride, Chlormequat chloride, Chlorocholine chloride, Cycocel, Cycocel Extra, Cycogan, Cysocel, Halloween, Lihocin, Microcil, New 5C Cycocel, New '5c' Cycocel, Retacel, Stabilan, TUR, Trimethyl-β-chloroethylammonium chloride, Tuval, WR 62, Zar
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCCl

Specifications

Quantity 100 g
InChI Formula InChI=1S/C5H13ClN.ClH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1H/q+1;/p-1
Formula Weight 157.0425
Chemical Name or Material Chlormequat chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.