missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(3,5-dimethylphenyl)phosphine, tech. 90%

Product Code. 11445630
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11445630

Brand: Thermo Scientific Alfa Aesar H25885.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74289-57-9 | C16H18ClP | 276.744 g/mol

Chlorobis(3,5-dimethylphenyl)phosphine is used in the synthesis of (S)- and (R)-2,2?-Bis[bis(3,5-dimethylphenyl)phosphinoyl]-5,5?,6,6?,7,7?,8,8?-octahydro-1,1?-binaphthyl (Xyl-H 8 -BINAPO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis(3,5-dimethylphenyl)phosphine is used in the synthesis of (S)- and (R)-2,2′-Bis[bis(3,5-dimethylphenyl)phosphinoyl]-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-binaphthyl (Xyl-H 8 -BINAPO).

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74289-57-9
Molecular Formula C16H18ClP
Molecular Weight (g/mol) 276.744
MDL Number MFCD01630841
InChI Key FCEBDAANWYNQMO-UHFFFAOYSA-N
Synonym bis 3,5-dimethylphenyl chlorophosphine, chlorobis 3,5-dimethylphenyl phosphine, chlorobis 3,5-dimethylphenyl phosphane, chloro-bis 3,5-dimethylphenyl phosphane, bis 3,5-dimethylphenyl phosphinous chloride, phosphinous chloride, bis 3,5-dimethylphenyl, bis 3,5-dimethylphenyl chlorophosphine, tech, phosphinous chloride,p,p-bis 3,5-dimethylphenyl
PubChem CID 4187520
IUPAC Name chloro-bis(3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1)P(C2=CC(=CC(=C2)C)C)Cl)C

Specifications

Quantity 5 g
UN Number UN3261
Solubility Information Reacts with water.
Formula Weight 276.75
Percent Purity 90%
Grade Technical
Chemical Name or Material Chlorobis(3,5-dimethylphenyl)phosphine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.