missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, TRC

Product Code. 30135529
Change view
Click to view available options
Quantity:
5 g
50 g
500 g
Unit Size:
500g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135529 500 g 500g
30135714 50 g 50g
17924105 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135529 Supplier Toronto Research Chemicals Supplier No. TRCC364530500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6 H5 Cl
Molecular Weight (g/mol) 112.56
Synonym Chlorobenzene, CP 27, Chlorobenezene, IP Carrier T 40, MCB, Monochlorobenzene, NSC 8433, Phenyl chloride, Tetrosin SP
IUPAC Name chlorobenzene
SMILES Clc1ccccc1

Specifications

Quantity 500 g
InChI Formula InChI=1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H
Formula Weight 112.008
Percent Purity >95%
Chemical Name or Material Chlorobenzene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.