missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, Puriss. p.a., ACS reagent, ≥99.5% (GC), Honeywell Riedel-de Haen™

Product Code. 16615773
Change view
Click to view available options
220 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16615773 220 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615773 Supplier Honeywell Riedel-de Haën Supplier No. 23570220KG
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point -45°C
CAS 108-90-7
Color Colorless
CAS Max % 1
Density 1.107 - 1.109 g/cm3 (20°C)
Quantity 220 kg
Flash Point 28°C
Molecular Formula C6H5Cl
Linear Formula C6H5Cl
Beilstein 605632
Boiling Point 130°C to 132°C (1013 hPa)
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILES ClC1=CC=CC=C1
IUPAC Name chlorobenzene
Molecular Weight (g/mol) 112.56
Grade ACS
Chemical Name or Material Chlorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.