missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals

Product Code. 11439940
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439940 1 g 1g
11449940 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11439940 Supplier Thermo Scientific Alfa Aesar Supplier No. 044712.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chlorobenzene-d5 is a halogenated D5 chlorobenzene for proteomics research. It is a precursor to deuterated dyes and rubbers. It is used as a high-boiling solvent in industrial applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3114-55-4
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 117.587
MDL Number MFCD00000531
InChI Key MVPPADPHJFYWMZ-RALIUCGRSA-N
Synonym chlorobenzene-d5, chloro 2h5 benzene, 1-chloro 2 h? benzene, chlorobenzene d5, benzene-d5, chloro, 2h5 phenyl chloride, 1-chloro 2h5 benzene, benzene-d5-, chloro, chlorobenzene, perdeutero, chlorobenzene-d5, 99 atom % d
PubChem CID 575975
IUPAC Name 1-chloro-2,3,4,5,6-pentadeuteriobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Physical Form Liquid
Density 1.157g/mL
Flash Point 24°C (75°F)
Odor Aromatic
Refractive Index 1.522
Quantity 1 g
Boiling Point 130°C
UN Number UN1134
Beilstein 1866335
Formula Weight 117.6
Percent Purity 99%
Chemical Name or Material Chlorobenzene-d5, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.