missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, ACS, 99.5%

Product Code. p-7050293
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 11455170

Brand: Thermo Scientific Alfa Aesar 036401.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol

Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45°C
Color Colorless
Density 1.106 g/mL
Boiling Point 132°C
Quantity 500 mL
Flash Point 28°C (82°F)
Odor Aromatic
Physical Form Liquid
Refractive Index 1.524
UN Number UN1134
Beilstein 605632
Merck Index 14,2121
Solubility Information Miscible with methanol and diethyl ether. Immiscible with water.
Formula Weight 112.56
Percent Purity 99.5%
Grade ACS
Chemical Name or Material Chlorobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.