missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene and Acetic Acid solution, Honeywell Burdick & Jackson™

Product Code. 16645913
Click to view available options
:
200 L
This item is not returnable. View return policy

Product Code. 16645913

Brand: Honeywell Burdick & Jackson 66163200L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Chemical Name or Material Chlorobenzene and Acetic Acid solution
CAS 108-90-7
Color Clear
Density 1.079 g/cm3 (20°C)
Boiling Point 118°C
Flash Point 27°C
Quantity 200 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.