missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetyl isocyanate, 90%, Tech.

Product Code. 10732885 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10732885 1 g Glass bottle 1g
10013680 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10732885 Supplier Thermo Scientific Acros Supplier No. 314300010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4461-30-7 | C3H2ClNO2 | 119.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4461-30-7
Molecular Formula C3H2ClNO2
Molecular Weight (g/mol) 119.51
MDL Number MFCD00002034
InChI Key MOVMEFHWBOWMFU-UHFFFAOYSA-N
Synonym chloroacetyl isocyanate, acetyl isocyanate, chloro, unii-h5wf2amh7j, chloroacetyl isocyante, h5wf2amh7j, snxhcfhai zyp@, chloroacetylisocyanate, acmc-20alh0, chloro-acetyl isocyanate, alpha-chloroacetyl isocyanate
PubChem CID 78209
IUPAC Name 2-chloroacetyl isocyanate
SMILES C(C(=O)N=C=O)Cl

Specifications

Color Yellow
Density 1.4030g/mL
Boiling Point 50.0°C to 55.0°C (20.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula ClCH2CONCO
Quantity 1 g
Merck Index 15, 2117
Specific Gravity 1.403
Formula Weight 119.51
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Chloroacetyl isocyanate, tech., 90%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.