missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetyl chloride, 98%

Product Code. 11690212
Change view
Click to view available options
Quantity:
25 mL
250 mL
500 mL
Unit Size:
250mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11690212 250 mL 250mL
11491960 25 mL 25mL
11600222 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11690212 Supplier Thermo Scientific Alfa Aesar Supplier No. A15846.AK

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 79-04-9 | C2H2Cl2O | 112.937 g/mol

Chloroacetyl chloride is an intermediate in the production of alachlor and butachlor. It is used to prepare phenacyl chloride through Friedel-Crafts acylation of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetyl chloride is an intermediate in the production of alachlor and butachlor. It is used to prepare phenacyl chloride through Friedel-Crafts acylation of benzene.

Solubility
Miscible with ethyl ether, acetone, benzene and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and water. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-04-9
Molecular Formula C2H2Cl2O
Molecular Weight (g/mol) 112.937
MDL Number MFCD00000725
InChI Key VGCXGMAHQTYDJK-UHFFFAOYSA-N
Synonym chloroacetyl chloride, acetyl chloride, chloro, chloracetyl chloride, chloroacetic chloride, monochloroacetyl chloride, chloroacetic acid chloride, chloroacetylchloride, chlorure de chloracetyle, unii-k5uml06yuo, chlorid kyseliny chloroctove
PubChem CID 6577
ChEBI CHEBI:34624
IUPAC Name 2-chloroacetyl chloride
SMILES C(C(=O)Cl)Cl

Specifications

Melting Point -22°C
Density 1.417
Boiling Point 105°C to 106°C
Odor Pungent
Linear Formula ClCH2COCl
Refractive Index 1.453
Quantity 250 mL
UN Number UN1752
Beilstein 605439
Sensitivity Moisture sensitive
Merck Index 14,2067
Solubility Information Miscible with ethyl ether,acetone,benzene and carbon tetrachloride.
Formula Weight 112.94
Percent Purity 98%
Chemical Name or Material Chloroacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.