missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroacetyl chloride, 98%, Thermo Scientific Chemicals
12.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -22.0°C |
Color | Colorless to Yellow |
Density | 1.4200g/mL |
Boiling Point | 105.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C2H2Cl2O |
Linear Formula | ClCH2COCl |
Refractive Index | 1.4520 to 1.4540 |
Show More |
Safety and Handling
Product Identifier
- Chloroacetyl chloride
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 1
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H290-May be corrosive to metals.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'