missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetonitrile, 98+%

Product Code. 11435213
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435213 500 g 500g
11425213 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435213 Supplier Thermo Scientific Alfa Aesar Supplier No. A12825.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 107-14-2 | C2H2ClN | 75.495 g/mol

Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen's condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen′s condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-14-2
Molecular Formula C2H2ClN
Molecular Weight (g/mol) 75.495
MDL Number MFCD00001885
InChI Key RENMDAKOXSCIGH-UHFFFAOYSA-N
Synonym chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro
PubChem CID 7856
ChEBI CHEBI:82416
IUPAC Name 2-chloroacetonitrile
SMILES C(C#N)Cl

Specifications

Density 1.195
Boiling Point 124°C to 126°C
Flash Point 56°C (132°F)
Linear Formula ClCH2CN
Refractive Index 1.423
Quantity 500 g
UN Number UN2668
Beilstein 506028
Solubility Information Immiscible with water.
Formula Weight 75.5
Percent Purity ≥98%
Chemical Name or Material Chloroacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.