missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetone, 96%, stabilized

Product Code. 10255940 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
25mL
5 L
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10255940

Brand: Thermo Scientific Acros 173470010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 78-95-5 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-95-5
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00000936
InChI Key BULLHNJGPPOUOX-UHFFFAOYSA-N
Synonym chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone
PubChem CID 6571
ChEBI CHEBI:47220
IUPAC Name 1-chloropropan-2-one
SMILES CC(=O)CCl

Specifications

Melting Point -44.5°C
Color Brown-Yellow to Yellow
Density 1.1600g/mL
Boiling Point 119.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2COCH3
Refractive Index 1.4310 to 1.4330
Quantity 1 L
Beilstein 01, 653
Merck Index 15, 2114
Specific Gravity 1.16
Solubility Information Solubility in water: 124g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
Stabilizer 0.1% epoxidized soybean oil
Viscosity 1.11 mPa.s (25°C)
Formula Weight 92.52
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Chloroacetone, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloroacetone, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Flammable liquid Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.