missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetone, 95%, stab. with 0.1% epoxidized soybean oil

Product Code. 11490980
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11490980

Brand: Thermo Scientific Alfa Aesar A11922.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-95-5 | C3H5ClO | 92.52 g/mol

Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.

Solubility
Soluble in water (124g/L), ether and chloroform.

Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Do not store together with strong basic or oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-95-5
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00000936
InChI Key BULLHNJGPPOUOX-UHFFFAOYSA-N
ChEBI CHEBI:47220
SMILES CC(=O)CCl

Specifications

Melting Point -45°C
Density 1.161
pH 4.3
Boiling Point 118°C to 120°C
Flash Point 40°C (104°F)
Assay Percent Range 95%, Stabilized with 0.1% Epoxidized Soybean Oil
Odor Pungent
Linear Formula ClCH2COCH3
Refractive Index 1.434
Quantity 1000 g
UN Number UN1695
Beilstein 605369
Merck Index 14,2114
Synonym chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone
Solubility Information Soluble in water (124g/L),ether and chloroform.
IUPAC Name 1-chloropropan-2-one
PubChem CID 6571
Formula Weight 92.53
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Chloroacetone, Stabilized with 0.1% epoxidized soybean oil
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloroacetone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Chloroacetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.