missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic Acid, Certified AR for Analysis, Fisher Chemical

Product Code. 10233660 Shop All Fisher Chemical Products
Change view
Click to view available options
500g, Glass Jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10233660 500g, Glass Jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10233660 Supplier Fisher Chemical Supplier No. C/4400/53
Quantity Price
1 £174.08 / 500g
4 £156.67 / 500g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 79-11-8
Melting Point 62°C
Color White
pH lt; 1
Boiling Point 190°C
Packaging Glass Jar
Molecular Formula C2H3ClO2
MDL Number 2683
Quantity 500 g
Formula Weight 94.5g/mol
Physical Form Flaky Solid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloroacetic acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.