missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroacetamidine hydrochloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 97°C to 103°C |
Quantity | 1 g |
UN Number | UN3261 |
Beilstein | 3678354 |
Sensitivity | Hygroscopic |
Solubility Information | Insoluble in water. |
Formula Weight | 129 |
Percent Purity | 96% |
Chemical Name or Material | Chloroacetamidine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'