missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetaldehyde dimethyl acetal, 97%

Product Code. 10325790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10325790 250 mL Glass bottle 250mL
10224280 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10325790 Supplier Thermo Scientific Acros Supplier No. 108502500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 97-97-2 | C4H9ClO2 | 124.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-97-2
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00000948
InChI Key CRZJPEIBPQWDGJ-UHFFFAOYSA-N
Synonym chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane
PubChem CID 60986
IUPAC Name 2-chloro-1,1-dimethoxyethane
SMILES COC(CCl)OC

Specifications

Melting Point -73.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 128.0°C to 130.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClCH2CH(OCH3)2
Refractive Index 1.4140 to 1.4160
Quantity 250 mL
Specific Gravity 1.09
Solubility Information Solubility in water: 52g/L (20 c).
Formula Weight 124.57
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chloroacetaldehyde dimethyl acetal, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroacetaldehyde dimethyl acetal
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.